
α,α-Dimethyl-4-fluorophenethylamine hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2413-54-9
:
Formula:C₁₀H₁₄FN·ClH
MDL Number:MFCD00075463
:203.684 g/mol
Melting Pt:180 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together