3-Methoxycarbonyl-5-trifluoromethoxylphenylboronic acid, pinacol ester 95%
Catalog # APOSPC412508-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:346.107 g/mol
Storage Temperature:Ambient
CAS Number:1150561-63-9
Formula:C₁₅H₁₈BF₃O₅
MDL Number:MFCD12026096