2,3-Difluoro-4-(methoxycarbonyl)phenylboronic acid
Catalog # APOSPC412360-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:215.947 g/mol
Storage Temperature:Refrigerator
CAS Number:1402238-29-2
Formula:C₈H₇BF₂O₄
MDL Number:MFCD18434428