
3-(Difluoromethoxy)-4-methoxyaniline hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1431962-33-2
:
Formula:C₈H₉F₂NO₂·ClH
MDL Number:MFCD25371276
:225.62 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together