Order Entry
Czech Republic
ContactUsLinkComponent
1,1-Difluorospiro[2.5]octan-6-one
1,1-Difluorospiro[2.5]octan-6-one
Catalog # APOSPC411288-1G
Supplier:  Apollo Scientific
CAS Number:  
1,1-Difluorospiro[2.5]octan-6-one
Catalog # APOSPC411288-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :160.161 g/mol
Storage Temperature:Ambient
CAS Number:1513853-81-0
Formula:C₈H₁₀F₂O
MDL Number:MFCD28126838