cis-3-Fluorocyclobutane-1-carboxylic acid
Catalog # APOSPC411271-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:118.106 g/mol
Storage Temperature:Ambient
CAS Number:123812-78-2
Formula:C₅H₇FO₂
MDL Number:MFCD16661179