1-(2,2-Difluoroethyl)-5-methyl-1H-pyrazol-3-amine
Catalog # APOSPC410498-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Typ balení:Bottle
- Bal.:1 g
- Sekvence:3047
Specifications
About this item
:
:161.153 g/mol
Storage Temperature:Ambient
CAS Number:1006486-89-0
Formula:C₆H₉F₂N₃
MDL Number:MFCD08696549