Order Entry
Czech Republic
ContactUsLinkComponent
1-(2,2-Difluoroethyl)-1H-pyrazole-3-carboxylic acid
Irritant
1-(2,2-Difluoroethyl)-1H-pyrazole-3-carboxylic acid
Catalog # APOSPC410494-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1-(2,2-Difluoroethyl)-1H-pyrazole-3-carboxylic acid
Catalog # APOSPC410494-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :176.121 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1006486-39-0
Formula:C₆H₆F₂N₂O₂
MDL Number:MFCD08696509