Order Entry
Czech Republic
ContactUsLinkComponent
2-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]ethanamine
2-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]ethanamine
Catalog # APOSPC410489-1G
Supplier:  Apollo Scientific
CAS Number:  
2-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]ethanamine
Catalog # APOSPC410489-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :219.207 g/mol
Storage Temperature:Ambient
CAS Number:1006479-50-0
Formula:C₉H₁₂F₃N₃
MDL Number:MFCD06804826