2-[3-(Trifluoromethyl)-1H-pyrazol-1-yl]butanoic acid
Catalog # APOSPC410479-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:222.164 g/mol
Storage Temperature:Ambient
CAS Number:1006473-54-6
Formula:C₈H₉F₃N₂O₂
MDL Number:MFCD04970063