3-Fluoro-4-[3-(trifluoromethyl)-1H-pyrazol-1-yl]aniline
Catalog # APOSPC410472-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:245.176 g/mol
Storage Temperature:Ambient
CAS Number:1006468-65-0
Formula:C₁₀H₇F₄N₃
MDL Number:MFCD08699601