Order Entry
Czech Republic
ContactUsLinkComponent
1-(2,2-Difluoroethyl)-1H-pyrazol-3-amine
1-(2,2-Difluoroethyl)-1H-pyrazol-3-amine
Catalog # APOSPC410464-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(2,2-Difluoroethyl)-1H-pyrazol-3-amine
Catalog # APOSPC410464-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :147.126 g/mol
Storage Temperature:Ambient
CAS Number:1006462-38-9
Formula:C₅H₇F₂N₃
MDL Number:MFCD08696542