1-{[5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]methyl}-1H-pyrazole-3-carboxylic acid
Catalog # APOSPC410458-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:274.199 g/mol
Storage Temperature:Ambient
CAS Number:1006459-10-4
Formula:C₁₀H₉F₃N₄O₂
MDL Number:MFCD04967761