Order Entry
Czech Republic
ContactUsLinkComponent
1-(2,2-Difluoroethyl)-4-nitro-1H-pyrazole
1-(2,2-Difluoroethyl)-4-nitro-1H-pyrazole
Catalog # APOSPC410439-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(2,2-Difluoroethyl)-4-nitro-1H-pyrazole
Catalog # APOSPC410439-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :177.109 g/mol
Storage Temperature:Ambient
CAS Number:1006442-51-8
Formula:C₅H₅F₂N₃O₂
MDL Number:MFCD08556227