5-(1,3-Dimethyl-1H-pyrazol-4-yl)-7-(trifluoromethyl)-4,5,6,7-tetrahydropyrazolo[1,5-a]pyrimidine
Catalog # APOSPC410432-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:285.268 g/mol
Storage Temperature:Ambient
CAS Number:1006434-36-1
Formula:C₁₂H₁₄F₃N₅
MDL Number:MFCD04968855