Order Entry
Czech Republic
ContactUsLinkComponent
4-Nitro-1-(2,2,2-trifluoroethyl)-1H-pyrazole-3-carboxylic acid
4-Nitro-1-(2,2,2-trifluoroethyl)-1H-pyrazole-3-carboxylic acid
Catalog # APOSPC410431-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Nitro-1-(2,2,2-trifluoroethyl)-1H-pyrazole-3-carboxylic acid
Catalog # APOSPC410431-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :239.109 g/mol
Storage Temperature:Ambient
CAS Number:1006433-24-4
Formula:C₆H₄F₃N₃O₄
MDL Number:MFCD08696333