Order Entry
Czech Republic
ContactUsLinkComponent
3-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]-2-methylpropanenitrile
3-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]-2-methylpropanenitrile
Catalog # APOSPC410429-1G
Supplier:  Apollo Scientific
CAS Number:  
3-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]-2-methylpropanenitrile
Catalog # APOSPC410429-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :243.228 g/mol
Storage Temperature:Ambient
CAS Number:1006356-58-6
Formula:C₁₁H₁₂F₃N₃
MDL Number:MFCD06739345