Order Entry
Czech Republic
ContactUsLinkComponent
5-Methyl-1-{[5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]methyl}-1H-pyrazol-3-amine
5-Methyl-1-{[5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]methyl}-1H-pyrazol-3-amine
Catalog # APOSPC410419-1G
Supplier:  Apollo Scientific
CAS Number:  
5-Methyl-1-{[5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]methyl}-1H-pyrazol-3-amine
Catalog # APOSPC410419-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :259.231 g/mol
Storage Temperature:Ambient
CAS Number:1006349-20-7
Formula:C₁₀H₁₂F₃N₅
MDL Number:MFCD04967743