Order Entry
Czech Republic
ContactUsLinkComponent
 
[3-(Trifluoromethyl)-1H-pyrazol-1-yl]acetonitrile
Catalog # APOSPC410413-1G
Supplier:  Apollo Scientific
CAS Number:  
[3-(Trifluoromethyl)-1H-pyrazol-1-yl]acetonitrile
Catalog # APOSPC410413-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :175.111 g/mol
Storage Temperature:Ambient
CAS Number:1006348-71-5
Formula:C₆H₄F₃N₃
MDL Number:MFCD04969709