Order Entry
Czech Republic
ContactUsLinkComponent
2-[5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]butanoic acid
2-[5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]butanoic acid
Catalog # APOSPC410408-1G
Supplier:  Apollo Scientific
CAS Number:  
2-[5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]butanoic acid
Catalog # APOSPC410408-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :236.191 g/mol
Storage Temperature:Ambient
CAS Number:1006348-57-7
Formula:C₉H₁₁F₃N₂O₂
MDL Number:MFCD04967809