Order Entry
Czech Republic
ContactUsLinkComponent
5-(1-Ethyl-3-methyl-1H-pyrazol-4-yl)-7-(trifluoromethyl)-4,5,6,7-tetrahydropyrazolo[1,5-a]pyrimidine
5-(1-Ethyl-3-methyl-1H-pyrazol-4-yl)-7-(trifluoromethyl)-4,5,6,7-tetrahydropyrazolo[1,5-a]pyrimidine
Catalog # APOSPC410403-1G
Supplier:  Apollo Scientific
CAS Number:  
5-(1-Ethyl-3-methyl-1H-pyrazol-4-yl)-7-(trifluoromethyl)-4,5,6,7-tetrahydropyrazolo[1,5-a]pyrimidine
Catalog # APOSPC410403-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :299.295 g/mol
Storage Temperature:Ambient
CAS Number:1006342-77-3
Formula:C₁₃H₁₆F₃N₅
MDL Number:MFCD04968858