Order Entry
Czech Republic
ContactUsLinkComponent
3-[5-Phenyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanenitrile
3-[5-Phenyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanenitrile
Catalog # APOSPC410390-1G
Supplier:  Apollo Scientific
CAS Number:  
3-[5-Phenyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanenitrile
Catalog # APOSPC410390-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :265.234 g/mol
Storage Temperature:Ambient
CAS Number:1006334-26-4
Formula:C₁₃H₁₀F₃N₃
MDL Number:MFCD08558312