Order Entry
Czech Republic
ContactUsLinkComponent
2-[4-Chloro-3-(trifluoromethyl)-1H-pyrazol-1-yl]butanoic acid
2-[4-Chloro-3-(trifluoromethyl)-1H-pyrazol-1-yl]butanoic acid
Catalog # APOSPC410383-1G
Supplier:  Apollo Scientific
CAS Number:  
2-[4-Chloro-3-(trifluoromethyl)-1H-pyrazol-1-yl]butanoic acid
Catalog # APOSPC410383-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :256.61 g/mol
Storage Temperature:Ambient
CAS Number:1006333-14-7
Formula:C₈H₈ClF₃N₂O₂
MDL Number:MFCD04970064