Order Entry
Czech Republic
ContactUsLinkComponent
1-[3-(Trifluoromethyl)-1H-pyrazol-1-yl]propan-2-amine
1-[3-(Trifluoromethyl)-1H-pyrazol-1-yl]propan-2-amine
Catalog # APOSPC410381-1G
Supplier:  Apollo Scientific
CAS Number:  
1-[3-(Trifluoromethyl)-1H-pyrazol-1-yl]propan-2-amine
Catalog # APOSPC410381-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :193.17 g/mol
Storage Temperature:Ambient
CAS Number:1006328-56-8
Formula:C₇H₁₀F₃N₃
MDL Number:MFCD04970168