Order Entry
Czech Republic
ContactUsLinkComponent
3-[3-Cyclopropyl-5-(trifluoromethyl)-1H-pyrazol-1-yl]propanenitrile
3-[3-Cyclopropyl-5-(trifluoromethyl)-1H-pyrazol-1-yl]propanenitrile
Catalog # APOSPC410372-1G
Supplier:  Apollo Scientific
CAS Number:  
3-[3-Cyclopropyl-5-(trifluoromethyl)-1H-pyrazol-1-yl]propanenitrile
Catalog # APOSPC410372-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :229.202 g/mol
Storage Temperature:Ambient
CAS Number:1006319-95-4
Formula:C₁₀H₁₀F₃N₃
MDL Number:MFCD08558159