Order Entry
Czech Republic
ContactUsLinkComponent
2-[4-Chloro-5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanehydrazide
2-[4-Chloro-5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanehydrazide
Catalog # APOSPC410356-1G
Supplier:  Apollo Scientific
CAS Number:  
2-[4-Chloro-5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanehydrazide
Catalog # APOSPC410356-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :270.639 g/mol
Storage Temperature:Ambient
CAS Number:1005651-08-0
Formula:C₈H₁₀ClF₃N₄O
MDL Number:MFCD03419691