Order Entry
Czech Republic
ContactUsLinkComponent
2-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanehydrazide
2-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanehydrazide
Catalog # APOSPC410355-1G
Supplier:  Apollo Scientific
CAS Number:  
2-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanehydrazide
Catalog # APOSPC410355-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :262.232 g/mol
Storage Temperature:Ambient
CAS Number:1005650-83-8
Formula:C₁₀H₁₃F₃N₄O
MDL Number:MFCD03419748