Order Entry
Czech Republic
ContactUsLinkComponent
2-[3-(Difluoromethyl)-5-methyl-1H-pyrazol-1-yl]propanehydrazide
2-[3-(Difluoromethyl)-5-methyl-1H-pyrazol-1-yl]propanehydrazide
Catalog # APOSPC410348-1G
Supplier:  Apollo Scientific
CAS Number:  
2-[3-(Difluoromethyl)-5-methyl-1H-pyrazol-1-yl]propanehydrazide
Catalog # APOSPC410348-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :218.204 g/mol
Storage Temperature:Ambient
CAS Number:1005631-76-4
Formula:C₈H₁₂F₂N₄O
MDL Number:MFCD03419705