Order Entry
Czech Republic
ContactUsLinkComponent
4-(Difluoromethyl)-6-(1-ethyl-5-methyl-1H-pyrazol-4-yl)-2-hydrazinylpyrimidine
4-(Difluoromethyl)-6-(1-ethyl-5-methyl-1H-pyrazol-4-yl)-2-hydrazinylpyrimidine
Catalog # APOSPC410330-1G
Supplier:  Apollo Scientific
CAS Number:  
4-(Difluoromethyl)-6-(1-ethyl-5-methyl-1H-pyrazol-4-yl)-2-hydrazinylpyrimidine
Catalog # APOSPC410330-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :268.266 g/mol
Storage Temperature:Ambient
CAS Number:1004644-07-8
Formula:C₁₁H₁₄F₂N₆
MDL Number:MFCD04970388