Order Entry
Czech Republic
ContactUsLinkComponent
4-(Difluoromethyl)-6-(1,3-dimethyl-1H-pyrazol-4-yl)-2-hydrazinylpyrimidine
4-(Difluoromethyl)-6-(1,3-dimethyl-1H-pyrazol-4-yl)-2-hydrazinylpyrimidine
Catalog # APOSPC410329-1G
Supplier:  Apollo Scientific
CAS Number:  
4-(Difluoromethyl)-6-(1,3-dimethyl-1H-pyrazol-4-yl)-2-hydrazinylpyrimidine
Catalog # APOSPC410329-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :254.239 g/mol
Storage Temperature:Ambient
CAS Number:1004644-06-7
Formula:C₁₀H₁₂F₂N₆
MDL Number:MFCD04970384