Order Entry
Czech Republic
ContactUsLinkComponent
4-(1,3-Dimethyl-1H-pyrazol-4-yl)-2-hydrazinyl-6-(trifluoromethyl)pyrimidine
4-(1,3-Dimethyl-1H-pyrazol-4-yl)-2-hydrazinyl-6-(trifluoromethyl)pyrimidine
Catalog # APOSPC410327-1G
Supplier:  Apollo Scientific
CAS Number:  
4-(1,3-Dimethyl-1H-pyrazol-4-yl)-2-hydrazinyl-6-(trifluoromethyl)pyrimidine
Catalog # APOSPC410327-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :272.23 g/mol
Storage Temperature:Ambient
CAS Number:1004644-03-4
Formula:C₁₀H₁₁F₃N₆
MDL Number:MFCD04970347