Order Entry
Czech Republic
ContactUsLinkComponent
3-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanehydrazide
3-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanehydrazide
Catalog # APOSPC410283-1G
Supplier:  Apollo Scientific
CAS Number:  
3-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanehydrazide
Catalog # APOSPC410283-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :262.232 g/mol
Storage Temperature:Ambient
CAS Number:1001518-97-3
Formula:C₁₀H₁₃F₃N₄O
MDL Number:MFCD03419754