Order Entry
Czech Republic
ContactUsLinkComponent
1-(2-Fluorobenzyl)-3-nitro-1H-pyrazole-5-carboxylic acid
1-(2-Fluorobenzyl)-3-nitro-1H-pyrazole-5-carboxylic acid
Catalog # APOSPC410170-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(2-Fluorobenzyl)-3-nitro-1H-pyrazole-5-carboxylic acid
Catalog # APOSPC410170-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :265.197 g/mol
Storage Temperature:Ambient
Formula:C₁₁H₈FN₃O₄
MDL Number:MFCD06016178