
2-(Difluoromethoxy)-5-methylaniline hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1431962-90-1
:
Formula:C₈H₉F₂NO·ClH
MDL Number:MFCD25371272
:209.621 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together