Order Entry
Czech Republic
ContactUsLinkComponent
2-[4-Amino-3-(trifluoromethyl)-1H-pyrazol-1-yl]butanoic acid hydrochloride
2-[4-Amino-3-(trifluoromethyl)-1H-pyrazol-1-yl]butanoic acid hydrochloride
Catalog # APOSPC410108-1G
Supplier:  Apollo Scientific
CAS Number:  
2-[4-Amino-3-(trifluoromethyl)-1H-pyrazol-1-yl]butanoic acid hydrochloride
Catalog # APOSPC410108-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :273.64 g/mol
Storage Temperature:Ambient
Formula:C₈H₁₀F₃N₃O₂·ClH
MDL Number:MFCD25371175