7-(4-Fluorophenyl)pyrazolo[1,5-a]pyrimidine-2-carboxylic acid hydrochloride
Catalog # APOSPC410083-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:293.681 g/mol
Storage Temperature:Ambient
Formula:C₁₃H₈FN₃O₂·ClH
MDL Number:MFCD25371107