Order Entry
Czech Republic
ContactUsLinkComponent
1-(2-Amino-4-fluorophenyl)-1H-pyrazole-3-carboxylic acid hydrochloride
1-(2-Amino-4-fluorophenyl)-1H-pyrazole-3-carboxylic acid hydrochloride
Catalog # APOSPC410073-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(2-Amino-4-fluorophenyl)-1H-pyrazole-3-carboxylic acid hydrochloride
Catalog # APOSPC410073-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :257.649 g/mol
Storage Temperature:Ambient
Formula:C₁₀H₈FN₃O₂·ClH
MDL Number:MFCD25368938