4-(3-Fluorophenoxy)benzonitrile
Catalog # APOSPC410057-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:213.207 g/mol
Storage Temperature:Ambient
CAS Number:877149-18-3
Formula:C₁₃H₈FNO
MDL Number:MFCD08559040