3-Chloro-2-(4-nitro-1H-pyrazol-1-yl)-5-(trifluoromethyl)pyridine
Catalog # APOSPC410046-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:292.602 g/mol
Storage Temperature:Ambient
Formula:C₉H₄ClF₃N₄O₂
MDL Number:MFCD08556251