Order Entry
Czech Republic
ContactUsLinkComponent
 
2-[3-(Trifluoromethyl)-5,6-dihydrocyclopenta[c]pyrazol-1(4H)-yl]acetohydrazide
Catalog # APOSPC410041-1G
Supplier:  Apollo Scientific
CAS Number:  
2-[3-(Trifluoromethyl)-5,6-dihydrocyclopenta[c]pyrazol-1(4H)-yl]acetohydrazide
Catalog # APOSPC410041-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :248.205 g/mol
Storage Temperature:Ambient
Formula:C₉H₁₁F₃N₄O
MDL Number:MFCD08558326