Order Entry
Czech Republic
ContactUsLinkComponent
1-(4-Fluorophenyl)-3,6-dimethyl-1H-pyrazolo[3,4-b]pyridine-4-carbohydrazide
1-(4-Fluorophenyl)-3,6-dimethyl-1H-pyrazolo[3,4-b]pyridine-4-carbohydrazide
Catalog # APOSPC410036-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(4-Fluorophenyl)-3,6-dimethyl-1H-pyrazolo[3,4-b]pyridine-4-carbohydrazide
Catalog # APOSPC410036-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :299.303 g/mol
Storage Temperature:Ambient
Formula:C₁₅H₁₄FN₅O
MDL Number:MFCD08558225