1-(3-Fluorophenyl)-6-methoxy-2,3,4,9-tetrahydro-1H-b-carboline hydrochloride
Catalog # APOSPC410034-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:332.8 g/mol
Storage Temperature:Ambient
CAS Number:1158793-60-2
Formula:C₁₈H₁₇FN₂O·ClH
MDL Number:MFCD02763964