Order Entry
Czech Republic
ContactUsLinkComponent
1-(4-Fluorophenyl)-6-methoxy-2,3,4,9-tetrahydro-1H-b-carboline hydrochloride
1-(4-Fluorophenyl)-6-methoxy-2,3,4,9-tetrahydro-1H-b-carboline hydrochloride
Catalog # APOSPC410033-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(4-Fluorophenyl)-6-methoxy-2,3,4,9-tetrahydro-1H-b-carboline hydrochloride
Catalog # APOSPC410033-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :332.8 g/mol
Storage Temperature:Ambient
CAS Number:1431965-08-0
Formula:C₁₈H₁₇FN₂O·ClH
MDL Number:MFCD02763959