Order Entry
Czech Republic
ContactUsLinkComponent
1-[(2-Chloro-4-fluorophenoxy)methyl]-4-nitro-1H-pyrazole
1-[(2-Chloro-4-fluorophenoxy)methyl]-4-nitro-1H-pyrazole
Catalog # APOSPC410024-1G
Supplier:  Apollo Scientific
CAS Number:  
1-[(2-Chloro-4-fluorophenoxy)methyl]-4-nitro-1H-pyrazole
Catalog # APOSPC410024-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :271.632 g/mol
Storage Temperature:Ambient
CAS Number:1003993-40-5
Formula:C₁₀H₇ClFN₃O₃
MDL Number:MFCD06740551