4-Nitro-1-(pentafluorobenzyl)-1H-pyrazole
Catalog # APOSPC410020-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:293.15 g/mol
Storage Temperature:Ambient
Formula:C₁₀H₄F₅N₃O₂
MDL Number:MFCD05861683