1-(2-Fluorobenzyl)-4-nitro-1H-pyrazole
Catalog # APOSPC410018-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:221.188 g/mol
Storage Temperature:Ambient
Formula:C₁₀H₈FN₃O₂
MDL Number:MFCD02732840