Order Entry
Czech Republic
ContactUsLinkComponent
4-Fluoro-4-methylpiperidine hydrochloride
Irritant
4-Fluoro-4-methylpiperidine hydrochloride
Catalog # APOSPC409056-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-Fluoro-4-methylpiperidine hydrochloride
Catalog # APOSPC409056-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :153.626 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1023305-87-4
Formula:C₆H₁₂FN·ClH
MDL Number:MFCD23381135