
N-BOC-cis-4-Trifluoromethyl-L-proline
Catalog # APOSPC409016-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Typ balení:Bottle
- Bal.:1 g
- Sekvence:3047
Specifications
About this item
:
:283.244 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:470482-41-8
Formula:C₁₁H₁₆F₃NO₄
MDL Number:MFCD08460520