Order Entry
Czech Republic
ContactUsLinkComponent
6-(Pentafluoroethyl)pyridine-3-carbaldehyde
6-(Pentafluoroethyl)pyridine-3-carbaldehyde
Catalog # APOSPC408847-1G
Supplier:  Apollo Scientific
CAS Number:  
6-(Pentafluoroethyl)pyridine-3-carbaldehyde
Catalog # APOSPC408847-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :225.115 g/mol
Storage Temperature:Ambient
CAS Number:1816282-87-7
Formula:C₈H₄F₅NO
MDL Number:MFCD28122895