Order Entry
Czech Republic
ContactUsLinkComponent
tert-Butyl 5-fluorospiro[indoline-3,4'-piperidine]-1'-carboxylate 96%
tert-Butyl 5-fluorospiro[indoline-3,4'-piperidine]-1'-carboxylate 96%
Catalog # APOSPC408429-250MG
Supplier:  Apollo Scientific
CAS Number:  
tert-Butyl 5-fluorospiro[indoline-3,4'-piperidine]-1'-carboxylate 96%
Catalog # APOSPC408429-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :306.375 g/mol
Storage Temperature:Ambient
CAS Number:167484-91-5
Formula:C₁₇H₂₃FN₂O₂
MDL Number:MFCD12198573